Learn More
Invitrogen™ UHMK1 Monoclonal Antibody (OTI2H4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525240
This item is not returnable.
View return policy
Description
UHMK1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Upon serum stimulation, KIS, a member of the Ser/Thr protein kinase family, phosphorylates CDKN1B/p27Kip1, thus controlling CDKN1B subcellular location and cell cycle progression in G1 phase. This protein, which contains 1 RRM (RNA recognition motif)domain, may be involved in trafficking and/or processing of RNA. KIS is widely expressed, with highest levels in skeletal muscle, kidney, placenta and peripheral blood leukocytes.
Specifications
| UHMK1 | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TAS1 | |
| UHMK1 | |
| Full length human recombinant protein of UHMK1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2H4 | |
| Unconjugated | |
| UHMK1 | |
| 4732477C12Rik; 4930500M09Rik; AA673513; AI449218; AU021979; kinase interacting stathmin; kinase interacting with leukemia-associated gene (stathmin); kinase interacting with stathmin; KIS; KIS protein kinase; KIST; PAM COOH-terminal interactor protein 2; P-CIP2; Serine/threonine-protein kinase Kist; U2AF homology motif (UHM) kinase 1; U2AF homology motif kinase 1; Uhmk1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 127933 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |