Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 Right-Hinged
Shop All asecos Products
Click to view available options
Configuration:
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump
3 Tray Shevles, 1 Perforated Insert, 1 Bottom Collecting Sump
4 x drawer
6 x drawer
Color:
Light Gray
Warning Yellow
Yellow
Depth (Metric) Exterior:
616 mm
749 mm
Unit Size:
Each
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding door: less required space in front of the cabinet while door is open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 6 x drawer |
| Depth (Metric) Exterior | 616 mm |
| Depth (Metric) Interior | 524 mm |
| Dimensions (D x W x H) Interior | 524 x 450 x 1742 mm |
| Dimensions (D x W x H) | 616 x 596 x 1968 mm |
| Door Style | Folding, Right-hinged |
| For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1968 mm |
| Height (Metric) Interior | 1742 mm |
| Show More |