Learn More
HTR5B (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577751
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
SR-5B, also designated 5-hydroxytryptamine (serotonin) receptor 5B (5-HT5B) and G protein-coupled receptor 134 (GPR134), is a receptor for the monoamine ligand serotonin (5-hydroxytryptamine, 5-HT). Serotonin is a neurotransmitter derived from serotonergic neurons in the central nervous system and enterochromaffin cells in the gastrointestinal tract. Serotonin actions are mediated by receptors that influence the biochemistry of depression, anxiety, sexuality and appetite. Rat SR-5B is present in serotonergic neurons in dorsal raphe (DR) and central superior nucleus (CS, median raphe nucleus). DR cell bodies showing SR-5B mRNA expression are abundant in the medial portions of the nucleus. CS coexpression of SR-5B receptor mRNA with serotonin transporter mRNA is high in the intermediate portions of the nucleus.
Specifications
| HTR5B (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Htr5b | |
| 5-HT5b, MR22, Serotonin receptor 5B, Htr5b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15564, 79247 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.85 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P31387, P35365 | |
| Htr5b | |
| Peptide CSDSPSSGRSMG, corresponding to amino acid residues 22-33 of rat 5-hydroxytryptamine receptor 5B | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |