Learn More
Invitrogen™ FAM69B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565871
This item is not returnable.
View return policy
Description
Immunogen sequence: EELGTQLRTCT TLSGLASQVE AHHSLVLSHL KTLLWK Highest antigen sequence identity to the following orthologs - mouse 92%, rat 95%.
This gene encodes a member of the FAM69 family of cysteine-rich type II transmembrane proteins. These proteins localize to the endoplasmic reticulum but their specific functions are unknown. [provided by RefSeq, Nov 2011].
Specifications
| FAM69B | |
| Polyclonal | |
| Unconjugated | |
| FAM69B | |
| AB030186; AW209000; B230317C12Rik; C9orf136; DIPK1B; Divergent protein kinase domain 1B; Fam69b; family with sequence similarity 69 member B; family with sequence similarity 69, member B; pancreatitis-induced protein 49; PIP49; pp6977; Protein FAM69B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138311 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VUD6 | |
| FAM69B | |
| Recombinant Human FAM69B. Recombinant protein control fragment (Product #RP-106542). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |