Learn More
Invitrogen™ Syntaxin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565209
This item is not returnable.
View return policy
Description
Immunogen sequence: DRLPDLTACRK NDDGDTVVVV EKDHFMDDFF HQVEEIRNTI DKITQYVEEV KK Highest antigen sequence identity to the following orthologs - mouse 80%, rat 75%.
The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Syntaxin 2 | |
| Polyclonal | |
| Unconjugated | |
| Stx2 | |
| AW538950; EPIM; Epimorphin; EPM; G1-536-1; I79_024396; MGC51014; repro34; Stx2; STX2A; STX2B; STX2C; Syn-2; syntaxin 2; syntaxin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2054 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P32856 | |
| Stx2 | |
| Recombinant Human STX2. Recombinant protein control fragment (Product #RP-105947). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |