Learn More
Invitrogen™ NIT1 Monoclonal Antibody (OTI4D12)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525068
This item is not returnable.
View return policy
Description
NIT1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.
Specifications
| NIT1 | |
| Monoclonal | |
| 0.66 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86X76 | |
| NIT1 | |
| Full length human recombinant protein of NIT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4D12 | |
| Unconjugated | |
| NIT1 | |
| AI255805; Deaminated glutathione amidase; dGSH amidase; ESTM30; MGC57670; Nit1; nitrilase 1; nitrilase homolog 1; rCG_20134; W57327 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |