Learn More
Invitrogen™ GGNBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570727
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Horse: 86%; Human: 100%; Rabbit: 92%.
This gene is the ortholog of the mouse gametogenetin-binding protein 1 gene. In human, the open reading frame is disrupted by a nonsense mutation after 8-aa; consequently, this gene is currently considered to be a unitary pseudogene in human even though it is functional in other mammals.
Specifications
| GGNBP1 | |
| Polyclonal | |
| Unconjugated | |
| GGNBP1 | |
| gametogenetin binding protein 1; gametogenetin binding protein 1 (pseudogene); gametogenetin-binding protein 1; Ggnbp1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 449520 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5YKI7 | |
| GGNBP1 | |
| synthetic peptide directed towards the N-terminal region of human GGNBP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |