Learn More
Invitrogen™ CCDC106 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560281
This item is not returnable.
View return policy
Description
Immunogen sequence: KLKSMSRAFE HHRVDRNTVA LTTPIAELLI VAPEKLAEVG EFDPSKERLL EYSRRCFLAL DDETLKKVQA LKKSKLLLPI TYRFK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. CCDC106 was initially identified as a p53-interacting protein by yeast two-hybrid screening. Other experiments demonstrated that CCDC106 co-localizes and interacts with p53 in the nucleus, inhibiting the transcriptional activity of p53 and stimulating p53 protein degradation, indicating that at least one of the functions of CCDC106 is acting as a negative regulator of p53.
Specifications
| CCDC106 | |
| Polyclonal | |
| Unconjugated | |
| CCDC106 | |
| BC018462; CCDC106; coiled-coil domain containing 106; coiled-coil domain-containing protein 106; HSU79303; MNCb-2071; protein predicted by clone 23882; RGD1566222; ZNF581 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29903 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BWC9 | |
| CCDC106 | |
| Recombinant protein corresponding to Human CCDC106. Recombinant protein control fragment (Product #RP-99161). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |