Learn More
Invitrogen™ DGKE Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA553558
Product Code. 15791491
  • £467.00 / 100µL
Description
Immunogen sequence: RDTDLFSQPT YCCVCAQHIL QGAFCDCCGL RVDEGCLRKA DKRFQCKEIM LKNDTKVLDA MPHHWIRGNV PLCSYCMVCK QQCGCQPKLC DYRCIWCQKT VHDECMKNSL KNEKCDFGEF KNLIIPPSYL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
Diacylglycerol (DAG) influences numerous cell signaling cascades by functioning as an intracellular, allosteric activator of protein kinase C (PKC), and as a potent activator of guanine nucleotide exchange factors. In order to maintain cellular homeostasis, intracellular DAG levels are tightly regulated by diacylglycerol kinases (DGKs, DAGKs), which phosphorylate DAG to phosphatidic acid, thus removing DAG. Human DGK-alpha (80 kDa), -beta (90 kDa), and - gamma (90 kDa) have calcium-binding EF-hand motifs at their N termini and are classified as type I DGKs. Human DGK- delta (130 kDa) and DGK-i (130 kDa) contain N-terminal pleckstrin homology (PH) domains and are classified as type II. Human DGK- epsilon (64 kDa) contains no identifiable regulatory domains and is classified as a type III DGK. Human DGK- zeta (104 kDa) and -iota (130 kDa) possess C-terminal ankyrin repeats and are classified as type IV DGKs. Human DGK-theta (110 kDa) contains 3 cysteine-rich domains and a PH domain and is classified as a type V DGK.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DGKE
Polyclonal
Unconjugated
Dgke
AHUS7; C87606; DAG kinase epsilon; DAGK5; DAGK6; DGK; DGKE; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon; diacylglycerol kinase, epsilon 64kDa; diglyceride kinase epsilon; EC 2.7.1.107; NPHS7
Rabbit
Antigen Affinity Chromatography
RUO
8526
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P52429
Dgke
Recombinant protein corresponding to Human DGKE. Recombinant protein control fragment (Product #RP-90372).