Learn More
Ohaus™ Explorer™ Analytical Balance
Product Code. 15776969 Shop All Ohaus Products
Click to view available options
Model:
EX224/AD
EX224M
Unit Size:
Each
Product Code. 15776969
Brand: Ohaus™ undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-performance, high-capacity, precise analytical scale with superior shock, spill, and splash resistance.
  • Intelligent performance
    • Optimized linearity and repeatability specifications and enhanced vibration filtering for better balance stability.
  • Intuitive operation
    • Large color touch-screen display, icon-driven application software that features 14 unique application modes, and a 3 level adjustable angle display make Explorer the easiest to use, most advanced balance in its class today.
  • Ingenious design
    • The base and display separate for maximum flexibility in any work area. Draftshield models isolate the weighing chamber to enhance stability and speed results while offering maximum accessibility to the weighing chamber through top-mounted side doors or through the versatile top door that flips or slides open. Hands-free entry is available with optional automated side doors.
  • Practical features
    • Explorer has up to four user-programmable touchless sensors to minimize cross contamination and wear and tear normally associated with repeated touching of the balance.
TRUSTED_SUSTAINABILITY
Specifications
Calibration AutoCal™ - Automatic
Connection Type RS232 (Included)USB (Included)Ethernet (Available as an Accessory)
Description Electronic Balance, EX224M/AD
Capacity 220 g
Display Type Full-color 5.7 in VGA, Touchscreen
Readability 0.100000 mg
Includes AC Adapter, RS232, USB, draftshield
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX)
Linearity 0.000200 g
Material ABS Top Housing, Stainless Steel Pan, Glass Draftshield
Model EX224/AD
Repeatability 0.000100 g
Stabilization Time 2.000 sec.
Dimensions (L x W x H) 393.00 x 230.00 x 350.000 mm
Length (English) 15.50 in.
Length (Metric) 393.00 mm
Width (English) 9.10 in.
Width (Metric) 230.00 mm
Width (Metric) Pan 90 mm
Height (English) 13.80 in.
Height (Metric) 350.000 mm
Depth (English) 15.50 in.
Depth (Metric) 393.00 mm