Learn More
MTCH1 Polyclonal Antibody
MTCH1 Polyclonal Antibody
Brand: Thermo Scientific PA553403
This item is not returnable.
View return policy
Description
MTCH1 Polyclonal Antibody for IHC
Specifications
| MTCH1 | |
| Polyclonal | |
| Unconjugated | |
| MTCH1 | |
| 2310034O17Rik; AI255158; AU018396; C77849; cell proliferation-inducing protein 60; CGI-64; mitochondrial carrier 1; mitochondrial carrier homolog 1; mitochondrial carrier homolog 1 (C. elegans); mitochondrial carrier-like protein 1; Mtch1; PIG60; presenilin-associated protein; PSAP; SLC25A49; solute carrier family 25, member 49; UNQ1871/PRO4314 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZJ7 | |
| MTCH1 | |
| Recombinant protein corresponding to Human MTCH1. Recombinant protein control fragment (Product #RP-90966). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |