Learn More
Invitrogen™ P4HA3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA552483
Product Code. 15770431
  • £467.00 / 100µL
Description
Immunogen sequence: TTPVANPLLA FTLIKRLQSD WRNVVHSLEA SENIRALKDG YEKVEQDLPA FEDLEGAARA LMRLQDVYML NVKGLARGVF QRVTGSAITD LYSPKRLFSL TGDDCFQVGK V Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P4HA3
Polyclonal
Unconjugated
P4ha3
4-PH alpha-3; collagen prolyl 4-hydroxylase alpha 3; collagen prolyl 4-hydroxylase alpha III subunit; collagen prolyl 4-hydroxylase alpha(III); C-P4H alpha III; D930031A02Rik; P4ha3; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide III; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-3; prolyl 4-hydroxylase subunit alpha 3; prolyl 4-hydroxylase subunit alpha-3; prolyl 4-hydroxylase, alpha polypeptide III; UNQ711/PRO1374
Rabbit
Antigen affinity chromatography
RUO
283208
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q7Z4N8
P4ha3
Recombinant protein corresponding to Human P4HA3. Recombinant protein control fragment (Product #RP-89693).