Learn More
Invitrogen™ POLR3K Polyclonal Antibody
GREENER_CHOICE
Product Code. 15760171 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15760171 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15760171 Supplier Invitrogen™ Supplier No. PA552220
Rabbit Polyclonal Antibody
Immunogen sequence: PGCGNGLIVE EGQRCHRFAC NTCPYVHNIT RKVTNRKYPK LKEVDDVLGG AAAWENVDST AESCPKCEHP RAYFMQLQTR SADEPMTTFY KCCNAQCGHR WRD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Eukaryotes produce 3 distinct classes of RNA polymerase, Pol I, II and III. Each polymerase is responsible for the synthesis of a different class of RNA. RNA polymerase III (Pol III) transcribes the 5S rRNA genes and all of the tRNA (transfer RNA) genes. POLR3K, also known as DNA-directed RNA polymerase III subunit RPC11, is a 108 amino acid nuclear protein that is one of the seventeen subunits that comprise Pol III. Specifically, POLR3K is a DNA-dependent RNA polymerase that uses the four ribonucleoside triphosphates to catalyze the transcription of DNA to RNA. The gene encoding POLR3K maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs and is associated with a variety of genetic disorders.
TRUSTED_SUSTAINABILITY
Specifications
Antigen POLR3K
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR3K
Gene Accession No. Q9Y2Y1
Gene Alias 12.3kDa; 1500004O14Rik; AI196849; AU014893; C11; C11-RNP3; DNA-directed RNA polymerase III subunit K; DNA-directed RNA polymerase III subunit RPC10; DNA-directed RNA polymerases III 12.5 kDa polypeptide; estrous-specific protein, 250 kDa; hRPC11; HsC11p; My010; POLR3K; polymerase (RNA) III (DNA directed) polypeptide K; polymerase (RNA) III (DNA directed) polypeptide K, 12.3 kDa; polymerase (RNA) III subunit K; RNA polymerase III 12.5 kDa subunit; RNA polymerase III subunit C10; RNA polymerase III subunit C11; RNA polymerase III subunit CII; RNA polymerase III subunit K; RPC10; RPC11; RPC12.5
Gene Symbols POLR3K
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR3K. Recombinant protein control fragment (Product #RP-89380).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG