Learn More
Invitrogen™ MVB12A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559772
This item is not returnable.
View return policy
Description
Immunogen sequence: LGATDTAVFD VRLSGKTKTV PGYLRIGDMG GFAIWCKKAK APRPVPKPRG LSRDMQGLSL DAASQPSKGG LLERTASRLG SRASTL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.
FAM125A, also named as MVB12A and CFBP, is a component of the ESCRT-I complex. FAM125A is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. FAM125A has three isoforms with MW 27-28 kDa and 25 kDa.
Specifications
| MVB12A | |
| Polyclonal | |
| Unconjugated | |
| MVB12A | |
| CFBP; CIN85/CD2AP family binding protein; CIN85/CD2AP family-binding protein; ESCRT-I complex subunit MVB12A; Fam125a; family with sequence similarity 125, member A; multivesicular body subunit 12A; Mvb12a; Protein FAM125A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EY5 | |
| MVB12A | |
| Recombinant protein corresponding to Human MVB12A. Recombinant protein control fragment (Product #RP-99121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |