Learn More
Invitrogen™ ISCA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560121
This item is not returnable.
View return policy
Description
Immunogen sequence: DKLSSEFVFN NPNIKGTCGC GESFNI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 96%.
Specifications
| ISCA1 | |
| Polyclonal | |
| Unconjugated | |
| ISCA1 | |
| 1810010A06Rik; GK004; HBLD2; HESB like domain containing 2; HesB protein; HESB-like domain-containing protein 2; hIscA; iron sulfur assembly protein IscA; iron-sulfur assembly protein IscA; iron-sulfur cluster assembly 1; iron-sulfur cluster assembly 1 homolog (S. cerevisiae); iron-sulfur cluster assembly 1 homolog, mitochondrial; ISA1; Isca1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUE6 | |
| ISCA1 | |
| Recombinant protein corresponding to Human ISCA1. Recombinant protein control fragment (Product #RP-100140). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |