Learn More
Invitrogen™ FTSJD2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA556317
Product Code. 15724231
  • £491.00 / 100µL
Description
Immunogen sequence: DDVRDYLFAV NIKLNQLRNT DSDVNLVVPL EVIKGDHEFT DYMIRSNESH CSLQIKALAK IHAFVQDTTL SEPRQAEIRK ECLRLWGIPD QARVAPSSSD PKS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.
S-adenosyl-L-methionine-dependent methyltransferase that mediates mRNA cap1 2'-O-ribose methylation to the 5'-cap structure of mRNAs. Methylates the ribose of the first nucleotide of a m(7)GpppG-capped mRNA and small nuclear RNA (snRNA) to produce m(7)GpppRm (cap1). Displays a preference for cap0 transcripts. Cap1 modification is linked to higher levels of translation. May be involved in the interferon response pathway. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FTSJD2
Polyclonal
Unconjugated
CMTR1
1300018I05Rik; AI451264; Cap methyltransferase 1; Cap1 2'O-ribose methyltransferase 1; cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; cmtr1; FtsJ methyltransferase domain containing 2; ftsJ methyltransferase domain-containing protein 2; ftsjd2; hMTr1; hypothetical protein LOC393102; interferon-stimulated gene 95 kDa protein; ISG95; KIAA0082; LOW QUALITY PROTEIN: cap-specific mRNA (nucleoside-2'-O-)-methyltransferase 1; LRRG00129; mKIAA0082; MTr1; RGD1307801; S-adenosyl-L-methionine-dependent methyltransferase ftsjd2; zgc:55336
Rabbit
Antigen affinity chromatography
RUO
23070, 309656, 74157
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.6 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5U2Z5, Q8N1G2, Q9DBC3
CMTR1
Recombinant protein corresponding to Human FTSJD2. Recombinant protein control fragment (Product #RP-94858).