Learn More
SMC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA563037
Product Code. 15720822
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Immunogen sequence: EVPSKRKNSA PQLPLLQSSG PFVEGSIVRI SMENFLTYDI CEVSPGPHLN MIVGANGTGK SSIVCAICLG LAGKPAFMGR ADKVGFFVKR GCSRGMVEIE LFRASGNLVI TR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Core component of the SMC5-SMC6 complex, a complex involved in DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMC5
Polyclonal
Unconjugated
SMC5
SMC5L1, SMC protein 5, SMC-5, SMC5 structural maintece of chromosomes 5-like 1, hSMC5, structural maintece of chromosomes protein 5
Rabbit
Antigen affinity chromatography
RUO
23137
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IY18
SMC5
Recombit protein corresponding to Human SMC5
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Research Use Only