Learn More
Thermo Scientific™ Depyrogenated Glass Containers
Product Code. 15669778
Change view
Click to view available options
Capacity (Metric):
4 mL
22 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Product Type:
Amber Packer Bottle
Container
Packer Bottle
Sample Jar
Vial
Wide Mouth Bottle
Color:
Amber
Clear
No. per Case:
6
12
24
72
Unit Size:
Pack of 12
Pack of 24
Pack of 6
Pack of 72
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Product Type Color No. per Case unitSize
15669778 22 mL Vial Clear 72 Pack of 72
16204651 4 mL Vial Clear 72 Pack of 72
15589475 60 mL Amber Packer Bottle Amber 24 Pack of 24
12721098 60 mL Sample Jar Clear 24 Pack of 24
15764022 125 mL Sample Jar Amber 24 Pack of 24
15874747 125 mL Sample Jar Clear 24 Pack of 24
15499014 125 mL Sample Jar Clear 24 Pack of 24
16401581 125 mL Packer Bottle Amber 24 Pack of 24
15763162 250 mL Sample Jar Amber 24 Pack of 24
15600709 250 mL Packer Bottle Amber 12 Pack of 12
16328657 250 mL Sample Jar Clear 24 Pack of 24
16338657 500 mL Sample Jar Clear 12 Pack of 12
15754022 1000 mL Container Amber 12 Pack of 12
15329541 2000 mL Wide Mouth Bottle Clear 6 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15669778 Supplier Thermo Scientific™ Supplier No. 13920C/CT/LP/PF
Only null left
Add to basket
Add to basket
Save on expensive capital equipment investments with ready-to-use glassware in a wide variety of sizes and configurations.
Save on expensive capital equipment investments with ready-to-use glassware in a wide variety of sizes and configurations. Thermo Scientific™ Depyrogenated Glass Containers are specially prepared to meet endotoxin levels of less than 0.06 EU/mL for use in terminal sterilization, water sampling and product storage.
  • Cleaning and packaging is performed in a Class 100/10 Cleanroom
  • Certificate of Analysis (COA) is included with each lot
Recommended for:
Endotoxin-free applications
Ordering Information:
Containers with additional low particle certification are available. Thermo Scientific also offers glass vials specially prepared to meet endotoxin levels of less than 0.06 EU/mL. Ask your customer service representative about our depyrogenation service with our validated process, performed in our Class 100 Cleanroom. 
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 22 mL
Capacity (English) 0.75 oz.
Color Clear
Diameter (English) 1.125 in.
Material Glass
No. per Case 72
Closure Color Black
Certifications/Compliance Meet endotoxin levels of less than 0.06 EU/mL; Includes certificate of analysis
Closure Included Yes
Mouth Narrow
Height (Metric) 57.15 mm
Product Type Vial
Height (English) 2.25 in.
Closure Liner PTFE/Silicone
Closure Material Polypropylene
Closure Size 24-400 mm
Diameter (Metric) 28.6 mm
Closure Type Closed Top