Learn More
CLEC1B Polyclonal Antibody
CLEC1B Polyclonal Antibody
Brand: Invitrogen PA550210
This item is not returnable.
View return policy
Description
CLEC1B Polyclonal Antibody for IHC (P)
Specifications
| CLEC1B | |
| Polyclonal | |
| Unconjugated | |
| CLEC1B | |
| 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51266 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9P126 | |
| CLEC1B | |
| Full length fusion protein of Human CLEC1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |