Learn More
Invitrogen™ Pleckstrin Monoclonal Antibody (2D8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519629
This item is not returnable.
View return policy
Description
Peptide Sequence: PETIDLGALY LSMKDTEKGI KELNLEKDKK IFNHCFTGNC VIDWLVSNQS VRNRQEGLMI ASSLLNEGYL QPAGDMSKSA VDGTAENPFL DNPDAFYYFP DSGFFCEENS.
Major protein kinase C substrate of platelets.
Specifications
| Pleckstrin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P08567 | |
| PLEK | |
| PLEK (AAH18549, 121 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 2D8 | |
| Unconjugated | |
| PLEK | |
| 2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5341 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |