Learn More
Invitrogen™ FAM160A2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA546454
Product Code. 15587172
  • £441.00 / 100µL
Description
Peptide sequence: AVPSAPTGPG PLLEFALHED LLTRVLTWQL QWDELGDGVE ERRAEQLKLF.
FAM160A2 is a component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FAM160A2
Polyclonal
Unconjugated
FAM160A2
C11orf56; FAM160A2; family with sequence similarity 160 member A2; family with sequence similarity 160, member A2; FHIP; FTS and Hook interacting protein; FTS and Hook-interacting protein; KIAA1759; Protein FAM160A2
Rabbit
Affinity chromatography
RUO
84067
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8N612
FAM160A2
Synthetic peptide directed towards the N-terminal of human FAM160A2 (aa 85-134).