Learn More
Invitrogen™ SETMAR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543676
This item is not returnable.
View return policy
Description
Peptide sequence: FLQGKRFHNQ QDAENAFQEF VESQSTDFYA TGINQLISRW QKCVDCNGSY Sequence homology: Cow: 100%; Human: 100%.
SETMAR (SET domain and mariner transposase fusion gene) is a histone methyltransferase that methylates Lys-4 and Lys-36 of histone H3, creating specific tags for epigenetic transcriptional activation. It specifically mediates dimethylation of H3 Lys-36. SETMAR has sequence-specific DNA-binding activity, and recognizes the 19-mer core of the 5'-terminal inverted repeats (TIRs) of the Hsmar1 transposable element. SETMAT has DNA nicking activity, and in vivo end-joining activity, and may mediate genomic integration of foreign DNA.
Specifications
| SETMAR | |
| Polyclonal | |
| Unconjugated | |
| SETMAR | |
| 5830404F24Rik; epidermal tumor expressed transcript 2; Etet2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Mar1; METNASE; SET domain and mariner transposase fusion gene; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain and mariner transposase fusion protein homolog; SET domain without mariner transposase fusion; SET domain without mariner transposase fusion protein; Setmar; Transposon Hsmar1 transposase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6419 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q53H47 | |
| SETMAR | |
| Synthetic peptide directed towards the C-terminal of human SETMAR (aa 633-682). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |