Learn More
Invitrogen™ TCP10L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542797
This item is not returnable.
View return policy
Description
Peptide sequence: ASPHAGQESH TLALEPAFGK ISPLSADEET IPKYAGHKNQ SATLLGQRSS Sequence homology: Human: 100%.
TCP10L is a human leucine zipper protein with transcription inhibition activity.
Specifications
| TCP10L | |
| Polyclonal | |
| Unconjugated | |
| TCP10L | |
| C21orf77; PRED77; t-complex 10 (a murine tcp homolog)-like; T-complex 10A-2; t-complex 10-like; T-complex protein 10A homolog 1; T-complex protein 10A homolog 2; T-complex protein 10A-1; T-complex protein 10A-2; TCP10A-2; TCP10L; TCP10-like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 140290 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TDR4 | |
| TCP10L | |
| Synthetic peptide directed towards the middle region of human TCP10L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |