Learn More
Invitrogen™ WFDC5 Monoclonal Antibody (5G5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523092
This item is not returnable.
View return policy
Description
Peptide Sequence: RKKGEKSGGC PPDDGPCLLS VPDQCVEDSQ CPLTRKCCYR ACFRQCVPRV SVKLGSCPED QLRCLSPMNH LCHKDSDCSG KKRCCHSACG RDCRDPARG.
WFDC5 is a putative acid-stable proteinase inhibitor.This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. Most WFDC proteins contain only one WFDC domain, and this encoded protein contains two WFDC domains. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.
Specifications
| WFDC5 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8TCV5 | |
| WFDC5 | |
| WFDC5 (NP_663627, 25 a.a. ~ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 5G5 | |
| Unconjugated | |
| WFDC5 | |
| BC019734; dJ211D12.5; p53-responsive gene 5 protein; Prg5; protease inhibitor WAP1; putative protease inhibitor WAP1; UNQ3036/PRO9842; WAP four-disulfide core domain 5; WAP four-disulfide core domain protein 5; WAP1; WFDC5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 149708 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |