Learn More
Phospho-IRF3 (Ser386) Polyclonal Antibody, Invitrogen™
Product Code. 15554551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15554551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15554551 Supplier Invitrogen Supplier No. PA537612
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rabbit Polyclonal Antibody
A suggested positive control for Western blot is 293 or COLO cells.
Phospho-IRF3 (Ser386) Polyclonal antibody specifically detects Phospho-IRF3 (Ser386) in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Phospho-IRF3 (Ser386)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IRF3
Gene Accession No. Q14653
Gene Alias C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3
Gene Symbols IRF3
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 386(A-S-S(p)-L-E) derived from Human IRF-3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3661
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG