Learn More
SNRK Polyclonal Antibody
SNRK Polyclonal Antibody
Brand: Invitrogen PA544539
This item is not returnable.
View return policy
Description
SNRK Polyclonal Antibody for Western Blot
Specifications
| SNRK | |
| Polyclonal | |
| Unconjugated | |
| SNRK | |
| 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRH2 | |
| SNRK | |
| Synthetic peptide directed towards the middle region of human SNRK. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |