Learn More
Invitrogen™ LONRF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542356
This item is not returnable.
View return policy
Description
Peptide sequence: MSSPAVARTS PGGSREMAPA PQGRGRFWEV GGGSGHRLER AAAESERWEL Sequence homology: Human: 100%; Mouse: 86%; Rat: 86%.
The function remains unknown.
Specifications
| LONRF1 | |
| Polyclonal | |
| Unconjugated | |
| LONRF1 | |
| LON peptidase N-terminal domain and ring finger 1; LON peptidase N-terminal domain and RING finger protein 1; LONRF1; RGD1562583; RING finger protein 191; RNF191 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 91694 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q17RB8 | |
| LONRF1 | |
| Synthetic peptide directed towards the N-terminal of human LONRF1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |