Learn More
Invitrogen™ SPAG11B Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA544106
Product Code. 15536352
  • £441.00 / 100µL
Description
Peptide sequence: MRQRLLPSVT SLLLVALLFP GSSQARHVNH SATEALGELR ERAPGQGTNG Sequence homology: Human: 100%.
SPAG11B is one of several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides.This gene encodes several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides. The gene is located on chromosome 8p23 near the defensin gene cluster. Alternative splicing of this gene results in seven transcript variants encoding different isoforms. Two different N-terminal and five different C-terminal protein sequences are encoded by the splice variants. Two additional variants have been described, but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPAG11B
Polyclonal
Unconjugated
SPAG11B
EDDM2B; EP2; EP2C; EP2D; epididymal protein 2B; HE2; HE2C; Human epididymis-specific protein 2; MGC61846; Protein EP2; SPAG11; SPAG11A; SPAG11B; Sperm antigen HE2; sperm associated antigen 11B; Sperm-associated antigen 11A; Sperm-associated antigen 11B
Rabbit
Affinity chromatography
RUO
10407
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q08648
SPAG11B
Synthetic peptide directed towards the N-terminal of human SPAG11B (aa 1-50).