Learn More
Invitrogen™ EXPH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542176
This item is not returnable.
View return policy
Description
Peptide sequence: QGRLWKPSFL KNPGFLKDDL RNPPNPSESL SSNSPSSQVP EDGLSPSEPL Sequence homology: Cow: 86%; Dog: 79%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 92%.
The function of this protein remains unknown.
Specifications
| EXPH5 | |
| Polyclonal | |
| Unconjugated | |
| EXPH5 | |
| exophilin 5; exophilin-5; EXPH5; KIAA0624; RGD1560308; SLAC2B; SLAC2-B; slp homolog lacking C2 domains b; synaptotagmin-like homologue lacking C2 domains b; synaptotagmin-like protein homolog lacking C2 domains b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NEV8 | |
| EXPH5 | |
| Synthetic peptide directed towards the middle region of human EXPH5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |