Learn More
Phospho-Cytokeratin 8 (Ser73) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA538072
Product Code. 15535001
  • £415.09 / 100µg
Description
Cytokeratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18. Cytokeratin 8 is primarily found in the non squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin polypeptides 8 and 18.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Cytokeratin 8 (Ser73)
Polyclonal
Unconjugated
KRT8
CK 8, CK8, CYK8, Cytokeratin endo A, K2C8, K8, KRT2-8, KRT8, Keratin, type II cytoskeletal 8, cytokeratin 8
Rabbit
Antigen affinity chromatography
RUO
16691, 3856
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P05787, P11679
KRT8
A synthetic phosphopeptide derived from human Keratin 8 around the phosphorylation site of Ser73 (L-L-SP-P-L)
100 μg
Primary
Human, Mouse