Learn More
Duplex 2209, 20 mesh woven from 0.3mm (0.012mm) dia wire, Thermo Scientific™
Product Code. 15467205
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
Unit Size:
Each
Product Code. 15467205
Brand: Thermo Scientific Alfa Aesar 046416.FL
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Duplex 2209
Name Note 20 mesh woven from 0.3mm (0.012mm) dia. wire
Quantity 75 x 75 mm
Mesh Size 20 Mesh
Odor Odorless
Form Wire Cloth