Learn More
Thermo Scientific™ Fibroblast Growth Factor 9, mouse
Click to view available options
Quantity:
10 μg
5 μg
Unit Size:
10µg
5µg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Molecular Weight (g/mol) | 23.3 kd |
| Quantity | 10 μg |
| Packaging | Plastic Bottle |
| Content And Storage | Store at -20°C |
| Form | Lyophilized |