Learn More
BTX™ Tweezertrode™ Electrode
Product Code. 15417330
Click to view available options
Diameter (Metric):
1 mm
3 mm
5 mm
7 mm
10 mm
Material:
Platinum
Stainless Steel
Unit Size:
Each
Product Code. 15417330
Brand: BTX™ 450489
Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.
  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned
Order Info
Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.
Specifications
For Use With (Application) in vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery
For Use With (Equipment) BTX 830, Gemini X2, ECM2001+, Legacy ECM 2001 Generator
Material Platinum
Product Line BTX Electroporation
Product Type Tweezer Style Electrode
Voltage 200 V
Description Platinum Tweezertrode, 5 mm Diameter
Diameter (English) 0.196 in.
Diameter (Metric) 5 mm
Electrode Shape Circular
Length (English) Electrode 0.47 in.
Length (Metric) Electrode 11.9 mm
Operating Temperature (English) 41°F to 104°F
Pulse Number Range 1 sec. to 99 msec.
Pulse Length Range 1 μsec. to 200 msec.
Operating Temperature (Metric) 5°C to 40°C
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (Metric) Gap Adjustable. <1 mm to >2 cm
Tip Type Tweezer Style
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less
Intended for Research use only. Not for use in diagnostic or clinical procedures.