Learn More
Invitrogen™ NPFF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533984
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey (94%) Marmoset (89%).
Receptor for NPAF (A-18-F-amide) and NPFF (F-8-F-amide) neuropeptides, also known as morphine-modulating peptides. Can also be activated by a variety of naturally occurring or synthetic FMRF-amide like ligands. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications
| NPFF2 | |
| Polyclonal | |
| Unconjugated | |
| NPFFR2 | |
| G protein-coupled receptor 74; Gpr74; G-protein coupled receptor 74; G-protein coupled receptor HLWAR77; HG31; HLWAR77; neuropeptide FF 2; neuropeptide FF receptor 2; Neuropeptide G-protein coupled receptor; neuropeptide NPFF receptor; Npff2; NPFF2 receptor; Npffr2; NPFF-R2; NPGP receptor; Npgpr | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y5X5 | |
| NPFFR2 | |
| Synthetic 18 amino acid peptide from C-terminus of human NPFFR2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |