Learn More
Invitrogen™ HTR1E Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533298
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Elephant, Panda, Bovine, Horse, Guinea pig, Turkey, Sparrow, Chicken, Lizard (100%); Bat, Opossum (94%); Stickleback, Zebrafish (88%). Antigen retrieval requires Proteinase K treatment.
G-protein coupled receptor for 5-hydroxytryptamine (serotonin). Also functions as a receptor for various alkaloids and psychoactive substances. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors, such as adenylate cyclase. Signaling inhibits adenylate cyclase activity.
Specifications
| HTR1E | |
| Polyclonal | |
| Unconjugated | |
| HTR1E | |
| 5-HT1E; 5-HT-1E; 5HT1E Receptor; 5-HT1e receptor; 5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled; 5-hydroxytryptamine receptor 1E; HT1e receptor; HTR1E; S31; serotonin receptor 1E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100065574, 100379275, 3354, 616356, 771973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P28566, Q6VB83 | |
| HTR1E | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human 5HT1E Receptor. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Bovine, Horse, Chicken, Guinea Pig | |
| Antibody | |
| IgG |