Learn More
Thermo Scientific Nunc Lab-Tek Sterile Petri Dishes, PS
Product Code. 15162944 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
100 mm
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15162944 100 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15162944 Supplier Thermo Scientific Nunc Lab-Tek Supplier No. 1400704
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These sterile and disposable polystyrene (PS) dishes are ideal for culturing fungi, bacteria, and other microorganisms.
  • Dishes work well in robotic dispensers due to flatness and uniform height.
  • Models 14007-10 and -11 are square in shape.
  • Models 14007-03 and -05 are 5 mm deeper and models 14007-04 and -14 are 10 mm deeper than standard dish depths to accommodate longer culture periods.
  • Sterilized by gamma irradiation.
Specifications
Color Transparent
Disposable Single-use
Material Polystyrene
Shape Square
Sterility Sterile
Type Petri Dish
For Use With (Application) For culturing fungi, bacteria, and other microorganisms
Quantity 300 Pk.
Height (Metric) 25 mm
Height (English) 1 in.
Diameter (English) 3.9 in.
Diameter (Metric) 100 mm