Learn More
ZIP14 Polyclonal Antibody
ZIP14 Polyclonal Antibody
Brand: Invitrogen PA533034
This item is not returnable.
View return policy
Description
ZIP14 Polyclonal Antibody for IHC (P)
Specifications
| ZIP14 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A14 | |
| cig19; Factor for adipocyte differentiation 123; Fad123; FAD-123; HMNDYT2; Kiaa0062; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; NET34; SLC39A14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; solute carrier family 39 member 14; Zinc transporter ZIP14; Zip14; ZIP-14; Zrt- and Cig19; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100152468, 23516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15043 | |
| SLC39A14 | |
| Synthetic 15 amino acid peptide from cytoplasmic domain of human SLC39A14. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Pig | |
| Antibody | |
| IgG |