Learn More
Invitrogen™ HN1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA521779
Product Code. 13289928
  • £441.00 / 100µL
Description
Recommended positive controls: 293T, A431, HeLa, K562, THP-1, HL-60. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HN1
Polyclonal
Unconjugated
JPT1
Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed
Rabbit
Antigen affinity chromatography
RUO
51155
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.22 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q9UK76
JPT1
Recombinant protein encompassing a sequence within the center region of human HN1. The exact sequence is proprietary.