Learn More
Invitrogen™ ZFYVE16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525044
This item is not returnable.
View return policy
Description
ZFYVE16 Polyclonal Antibody for Western Blot, IHC (P), Flow
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
Specifications
| ZFYVE16 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q7Z3T8 | |
| ZFYVE16 | |
| KLH conjugated synthetic peptide between 375-404 amino acids from the N-terminal region of human ZFYVE16. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| ZFYVE16 | |
| AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9765 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |