Få mere at vide
anti-Glucose Transporter GLUT3, Polyclonal, Abcam™
Brand: Abcam AB41525-100UG
Beskrivelse
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human
Immunogen: Synthetic peptide from the C terminus of Glucose Transporter GLUT3.
Applications:
Dot Blotting, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Tekniske data
| Glucose Transporter GLUT3 | |
| 0.5mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cancer and Tumor Biology, Cell Biology, Signal Transduction | |
| Human, Mouse, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide from the C terminus of Glucose Transporter GLUT3. | |
| 100 μg | |
| Primary | |
| IgG |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.