Learn More
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Popular threaded coned port configurations support quality high-pressure connections
Brand: Idex F-330X
Description
Popular threaded coned port configurations support quality high-pressure connections
- Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Specifications
| Fingertight Two-Piece Fitting | |
| PEEK | |
| 1/16 in. OD Tubing |
| Ferrule | |
| Natural |