Learn More
AZOWIPE™ Wipes
Product Code. 12899297
Click to view available options
Packaging Quantity:
200 Pack
2400 Pack
Unit Size:
6kg
Product Code. 12899297
Brand: Vernacare 81123
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bactericidal hard surface disinfectant wipes. AZOWIPE™ Wipes are convenient for use in all clinical environments and are an important part of a robust infection control protocol.
  • Made from a viscose rayon nonwoven material strong and durable in use
  • Foil sealed - prevents evaporation in storage
  • Suitable for all hard surfaces - one product from a single source
  • Contains NO preservative - no smearing or streaking when in use
  • Specially designed canister - easy to dispense wipes
  • 70% Solution of Isopropyl Alcohol BP
  • AZOWIPE - for the disinfection of hard surfaces
  • Dimensions: 200mm x 220mm
Specifications
Product Type Wipes
Dimensions (L x W) 220 x 200 mm
Packaging Quantity 2400 Pack