Learn More
Invitrogen™ C/EBP beta Monoclonal Antibody (3H9)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA515893
Product Code. 12847912
  • £484.00 / 100µL
Description
MA5-15893 targets CEBPB in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15893 immunogen is purified recombinant fragment of human CEBPB expressed in E. Coli. MA5-15893 detects CEBPB which has a predicted molecular weight of approximately 36kDa.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C/EBP beta
Monoclonal
Conc. Not Determined
Ascites with 0.03% sodium azide
P17676
Cebpb
Purified recombinant fragment of human CEBPB expressed in E. Coli.
RUO
1051
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Western Blot
3H9
Unconjugated
Cebpb
AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta