Learn More
Mettler Toledo™ Excellence XS Precision Balances
Product Code. 12740674 Shop All Mettler Toledo Products
Click to view available options
Capacity:
1.2, 6.1 kg
120, 610 g
2.1 kg
4.1 kg
6.1 kg
8.1 kg
Readability:
0.001 g
0.01 g
0.01, 0.1 g
0.1 g
100 mg
Stabilization Time:
0.8 sec.
1 sec.
1.2 sec.
1.5 sec.
Connection Type:
23/s, bluetooth (wireless), ethernet, PS/2, USB
Ethernet, BTS, BT, RS232C, PS/2, LC, MM, USB
RS-232C, bluetooth (wireless), ethernet, PS/2, USB
Unit Size:
Each
Product Code. 12740674
Brand: Mettler Toledo™ 11130171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for highest efficiency in everyday operation, reliabile and ease of use
  • Fast, precise weighing results
  • Touch-screen display with graphical user guidance
  • MonoBloc High-Speed weighing cell, accelerated via dedicated processor
  • Alphanumeric sample ID entry (up to three IDs per sample)
  • GLP/GMP-compliant printouts
  • Verified versions and stand-alone platforms available
Design
  • Large, square weighing pans with up to 30% more surface area
  • Draft shield (1mg models) for direct access to weighing pan from any side
  • Robust construction and IP54 in-use protection
Applications
Weighing, statistics, %-weighing, animal/dynamic weighing, piece counting, formulation and density determination
Warranty
One year standard warranty with return of warranty card. Warranty can be extended for 12 additional months by ordering Cat. No. 09-313-190 or extended 24 additional months by ordering Cat. No. 01-910-600.
TRUSTED_SUSTAINABILITY
Specifications
Automation Interface Touch screen
Certifications/Compliance GLP, GMP, ISO 9001, ISO 14001, CE
Connection Type 23/s, bluetooth (wireless), ethernet, PS/2, USB
Capacity 2.1 kg
Display Type Color Graphic
Readability 0.01 g
IP Rating IP54
Leveling Automatic
Linearity ±20 mg
Material Die-cast Aluminum, Plastic, Chrome Steel
Model XS2002S
Repeatability 8 mg
Stabilization Time 1.2 sec.
Test Load 1 kg
Type Precision Balance
Warranty 1 Year
Width (English) 14.4 in.
Width (English) Pan 6.6 in.
Width (Metric) 36.6 cm
Width (Metric) Pan 17 cm
Height (English) 3.7 in.
Height (Metric) 9.6 cm
Depth (English) 7.6 in.
Depth (English) Pan 8 in.
Depth (Metric) 19.4 cm
Depth (Metric) Pan 20.5 cm
Weight (English) 14.6 lb.
Weight (Metric) 6.6 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz