Learn More
GLUT-1 Rabbit Polyclonal Antibody, Epredia™
GLUT-1, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand:  Epredia RB-9052-P1
 View more versions of this product
Product Code. 12663417
  • ONLINE EXCLUSIVE PRICE
    £555.00
    £403.36 / 500µL
    Save £151.64
    27% Off
Description
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1
Molecular Weight: 55 kDa, 45 kDa depending on the extent of glycosylation
Positive Control: Rat brain lysate; Breast CA, squamous cell CA, nonsmall-cell lung CA and esophageal CA
Cellular Localization: Membrane and focal cytoplasmic
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLUT-1
Polyclonal
200µg/mL
Rabbit
500 μL
Steroids Hormones and Growth Factors
Human
IgG
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Unconjugated
A synthetic peptide derived from C-terminal of human GLUT-1
IVD
Primary
Concentrated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications