Learn More
Macherey-Nagel™ NucleoSpin™ Food, Mini kit for DNA from food
Description
Features:
- For rapid small-scale isolation of genomic DNA from food and feed
- Complete removal of PCR inhibitors – get high quality DNA
- Even low amounts of partially degraded DNA can be purified from complex matrices
- Fast and easy procedure DNA from various sample materials – highest flexibility
Applications:
- DNA from complex matrices, e.g., processed food, soy (milk and flour), chocolate, cereals, meat, animal feed
- Typical downstream applications: real-time PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
| Detection Method | Silica-membrane Technology |
| Format | Mini Spin Column |
| For Use With (Equipment) | PCR, Southern Blotting, Enzymatic Reactions |
| Includes | 2mL Collection Tubes, Buffers, Proteinase K |
| No. of Reactions | 50 Preps |
| Product Type | DNA Isolation Columns |
| Quantity | 1 Each |
| Sample Volume | <200 mg |
| Test Time | 30 min. |
| Yield | 0.1 to 10 μg (200 mg food) |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: guanidine hydrochloride/dodecyl sulfate,sodium salt/ethanol