Learn More
Shamrock™ Color Coded Multi-Purpose Labeling Tape, 0.75 in. (19 mm) Roll
Click to view available options
Color:
Aqua
Blue
Copper
Dark Brown
Gray
Green
Lavender
Lime Green
Magenta
Orange
Pink
Red
Rose
Tan
Violet
White
Yellow
Unit Size:
12m
Description
- Resistant to water, oil and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Specifications
| Autoclavable | Autoclavable |
| Adhesive Type | Removable |
| Color | Lime Green |
| Display | Blank |
| Format | Roll |
| Product Type | Color-coded Multi-purpose Labeling Tape |
| Sterility | Non-sterile |
| For Use With (Application) | Ideal for Universal Marking Pen, a Pencil and Felt |
| For Use With (Equipment) | Autoclave, Freezer, Fridge and Oven |
| Quantity | 1 Ea. |
| Show More |