Learn More
Invitrogen™ GPBB Monoclonal Antibody (1G6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA121528
This item is not returnable.
View return policy
Description
MA1-21528 detects GPBB in human samples.
The protein encoded by this gene is a glycogen phosphorylase found predominantly in the brain. The encoded protein forms homodimers which can associate into homotetramers, the enzymatically active form of glycogen phosphorylase. The activity of this enzyme is positively regulated by AMP and negatively regulated by ATP, ADP, and glucose-6-phosphate. This enzyme catalyzes the rate-determining step in glycogen degradation.
Specifications
| GPBB | |
| Monoclonal | |
| 5.8 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P11216 | |
| PYGB | |
| Human GPBB. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Affinity Purification, ELISA, Western Blot | |
| 1G6 | |
| Unconjugated | |
| PYGB | |
| brain glycogen phosphorylase; glycogen phosphorylase B; glycogen phosphorylase, brain form; GLYPHOA; GPBB; MGC9213; phosphorylase, glycogen; brain; PYGB | |
| Mouse | |
| Protein A | |
| RUO | |
| 5834 | |
| 4°C | |
| Liquid |