Learn More
Invitrogen™ Nidogen Monoclonal Antibody (ELM1)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA106501
Product Code. 11528812
  • £473.00 / 100µL
Description
MA1-06501 detects entactin in human samples. MA1-06501 has sucessfully been used in Western blotting, immunosytochemistry and immunohistochemistry. By Western blotting, it detects a ~150 kDa protein representing entactin. The MA1-06501 immunogen is partially purified preparation of laminin from the EHS mouse tumor. Store at 4°C or in small aliquots at -20°C.
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nidogen
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P10493
NID1
Partially purified preparation of laminin from the EHS mouse tumor.
100 μg
Primary
Mouse
Antibody
IgG2a
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
ELM1
Unconjugated
NID1
A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Rat
Purified
RUO
18073
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.