Learn More
Reagecon 100ML ICP STANDARD MERCURY 10% HNO3 1,000UG/ML
Brand: Reagecon PHG2A6
100ML ICP STANDARD MERCURY 10% HNO3 1,000UG/ML
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MERCURY ICP STANDARD SOLUTION
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, mercury